Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 68]
Catalog # ABNOMAB1341
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:immunodeficiency virus type 1 p24
- Clonality:Monoclonal
- Clone:68
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Cross adsorption:No
- Format:Lyophilized
- Storage buffer:PBS
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length HIV-1 p24.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus type 1 p24.
Type: Primary
Antigen: immunodeficiency virus type 1 p24
Clonality: Monoclonal
Clone: 68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: