Order Entry
Czech Republic
ContactUsLinkComponent
Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 49]
Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 49]
Catalog # ABNOMAB1342
Supplier:  Abnova
Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 49]
Catalog # ABNOMAB1342
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    immunodeficiency virus type 1 p24
  • Clonality:
    Monoclonal
  • Clone:
    49
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length HIV-1 p24.
  • Size:
    200 μg
  • Pk:
    200 µg

Specifications

About this item

Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus type 1 p24.

Type: Primary
Antigen: immunodeficiency virus type 1 p24
Clonality: Monoclonal
Clone: 49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: