Anti-COL3A1 Mouse Monoclonal Antibody [clone: COL-29]
ABNOMAB1514
: Abnova
- Antibody type:Primary
- Antigen name:Collagen, type III, alpha 1
- Antigen symbol:COL3A1
- Clonality:Monoclonal
- Clone:COL-29
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:1281
- Antigen synonyms:EDS4A
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human COL3A1.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against native COL3A1.
Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.5-1 ?g/ml; Immunohistochemistry (Frozen sections): 0.5-1 ?g/ml
Type: Primary
Antigen: COL3A1
Clonality: Monoclonal
Clone: COL-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat