Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: WK4]
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: WK4]
Catalog # ABNOMAB1721
Supplier:  Abnova
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: WK4]
Catalog # ABNOMAB1721
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 3 (amino acid transporter heavy chain), member 2
  • Antigen symbol:
    Solute carrier family 3, member 2
  • Clonality:
    Monoclonal
  • Clone:
    WK4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6520
  • Antigen synonyms:
    4F2HC|4F2|NACAE|CD98HC|MDU1|CD98|4T2HC
  • Storage buffer:
    PBS (0,1% proclin, 2.0% Block Ace)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified from human cancer cel line.
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Mouse monoclonal antibody raised against native SLC3A2.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/ml; Immunoprecipitation: 5-10 ?g/ml

Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: WK4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human