Anti-CML Monoclonal Antibody [clone: CMS-10]
Catalog # ABNOMAB1732
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CML
- Clonality:Monoclonal
- Clone:CMS-10
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1% proclin, 2.0% Block Ace)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CML conjugated with KLH.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse monoclonal antibody raised against CML.
Recommended Dilutions: Immunohistochemistry: 5-10 ?g/ml; ELISA: 0.1-1.0 ?g/ml
Type: Primary
Antigen: CML
Clonality: Monoclonal
Clone: CMS-10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human, Mouse, Rat