Anti-3-DG-imidazolone Monoclonal Antibody [clone: JNH 27]
Catalog # ABNOMAB1738
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:3-DG-imidazolone
- Antigen symbol:3-DG-imidazolone
- Clonality:Monoclonal
- Clone:JNH 27
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1% proclin, 2.0% Block Ace)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:3-DG-imidazolone conjugated with HSA.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse monoclonal antibody raised against 3-DG-imidazolone.
Recommended Dilutions: Immunohistochemistry: 7 ?g/ml
Type: Primary
Antigen: 3-DG-imidazolone
Clonality: Monoclonal
Clone: JNH 27
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human, Mouse, Rat