Anti-VPA Monoclonal Antibody [clone: 13-615.3]
Catalog # ABNOMAB2270
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Valproic Acid
- Antigen symbol:VPA
- Clonality:Monoclonal
- Clone:13-615.3
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG1
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Valproic acid conjugated with KLH.
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against Valproic acid.
Type: Primary
Antigen: VPA
Clonality: Monoclonal
Clone: 13-615.3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: