Anti-N-Acetylprocainamide Monoclonal Antibody [clone: 11-233.1.3]
ABNOMAB2277
: Abnova
- Antibody type:Primary
- Antigen name:N-acetylprocainamide
- Antigen symbol:NAPA
- Clonality:Monoclonal
- Clone:11-233.1.3
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG1
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:N-Acetylprocainamide conjugated with KLH.
- Pk:100 µG
Mouse monoclonal antibody raised against N-Acetylprocainamide.
Type: Primary
Antigen: N-Acetylprocainamide
Clonality: Monoclonal
Clone: 11-233.1.3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: