Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Catalog # ABNOMAB2355
Supplier:  Abnova
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Catalog # ABNOMAB2355
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromatin Assembly Factor 1, Subunit B (p60)
  • Antigen symbol:
    CHAF1B
  • Clonality:
    Monoclonal
  • Clone:
    SS 53
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8208
  • Antigen synonyms:
    CAF-1|MPHOSPH7|CAF1P60|CAF-IP60|MPP7|CAF1A|CAF1
  • Storage buffer:
    buffer containing 0,1% sodium azide
  • Storage temperature:
    Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human CHAF1B.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant CHAF1B.

Type: Primary
Antigen: CHAF1B
Clonality: Monoclonal
Clone: SS 53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human