Anti-Bu-1b Mouse Monoclonal Antibody [clone: 5-11G2]
Catalog # ABNOMAB3276
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Bu-1b
- Clonality:Monoclonal
- Clone:5-11G2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Chicken
- Cross adsorption:No
- Format:Liquid
- Storage buffer:100 mM BBS, pH 8,2
- Storage temperature:Store at 4 °C.
- Immunogen:Bursal cells from 2- to 5-week-old CHA (Bu-1b) strain chickens.
- Pk:500 µG
Specifications
About this item
Mouse monoclonal antibody raised against Bu-1b.
Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells
Type: Primary
Antigen: Bu-1b
Clonality: Monoclonal
Clone: 5-11G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken