Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-FANCD2 Monoclonal Antibody [clone: FI-17]
  ABNOMAB2398
 :  Abnova
Anti-FANCD2 Monoclonal Antibody [clone: FI-17]
  ABNOMAB2398
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Fanconi Anemia Complementation Group D2
  • Antigen symbol:
    FANCD2
  • Clonality:
    Monoclonal
  • Clone:
    FI-17
  • Conjugation:
    Unconjugated
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Format:
    Liquid
  • Antigen synonyms:
    FAD|FA4|FANCD|FACD|FA-D2|FAD2
  • Storage buffer:
    buffer containing 0,1% sodium azide
  • Storage temperature:
    Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant fusion protein corresponding to FANCD2.
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Mouse monoclonal antibody raised against recombinant FANCD2.

Recommended Dilutions: ChIP: 1:1000-1:2000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: FANCD2
Clonality: Monoclonal
Clone: FI-17
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human