Anti-CISH Rabbit Polyclonal Antibody
Catalog # ABNOPAB12758
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokine inducible SH2-Containing protein
- Antigen symbol:CISH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:BACTS2|G18|SOCS|CIS-1|CIS
- Storage buffer:PBS, pH 7,2 (10% Proclin300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal domain of human, mouse and rat CISH.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of CISH.
Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 0.5-2 ?g/ml
Type: Primary
Antigen: CISH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat