Anti-COL1A1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13714
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Collagen type I alpha 1
- Antigen symbol:COL1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:29393|84352
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with distilled water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified rat Col1a1/Col1a2.
- Size:1 mL
- Pk:1 mg
Specifications
About this item
Rabbit polyclonal antibody raised against native Col1a1/Col1a2.
Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:10-1:20
Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat