Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD59 Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-43]
  ABNOMAB5011
 :  Abnova
Anti-CD59 Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-43]
  ABNOMAB5011
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    CD59 Molecule, Complement Regulatory Protein
  • Antigen symbol:
    CD59
  • Clonality:
    Monoclonal
  • Clone:
    MEM-43
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    966
  • Antigen synonyms:
    MIRL|HRF-20|MEM43|1F5|MACIF|MIN3|EJ16|MSK21|EL32|HRF20|p18-20|MIC11|MIN1|16.3A5|MIN2|MAC-IP|G344|EJ30
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD59 from thymocytes and T lymphocytes.
  • Size:
    100 Reactions
  • Pk:
    100 Tests

 

 

Mouse monoclonal antibody raised against native CD59.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: PE (Phycoerythrin)
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human