Order Entry
Czech Republic
ContactUsLinkComponent
Anti-B2M Mouse Monoclonal Antibody (Biotin) [clone: B2M-01]
Anti-B2M Mouse Monoclonal Antibody (Biotin) [clone: B2M-01]
Catalog # ABNOMAB5020
Supplier:  Abnova
Anti-B2M Mouse Monoclonal Antibody (Biotin) [clone: B2M-01]
Catalog # ABNOMAB5020
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta-2-Microglobulin
  • Antigen symbol:
    B2M
  • Clonality:
    Monoclonal
  • Clone:
    B2M-01
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    567
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Purified human B2M.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against B2M.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: B2M-01
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human