Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-SLC3A2 Mouse Monoclonal Antibody (Biotin) [clone: MEM-108]
  ABNOMAB5040
 :  Abnova
Anti-SLC3A2 Mouse Monoclonal Antibody (Biotin) [clone: MEM-108]
  ABNOMAB5040
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 3 Member A2
  • Antigen symbol:
    Solute carrier family 3, member 2
  • Clonality:
    Monoclonal
  • Clone:
    MEM-108
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6520
  • Antigen synonyms:
    4F2HC|4F2|NACAE|CD98HC|MDU1|CD98|4T2HC
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified SLC3A2 from Raji Burkitt's lymphoma cell line.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against native SLC3A2.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: MEM-108
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human