Anti-C-peptide Mouse Monoclonal Antibody [clone: CII-29]
ABNOMAB2810
: Abnova
- Antibody type:Primary
- Antigen name:C-peptide
- Clonality:Monoclonal
- Clone:CII-29
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:A synthetic peptide corresponding to rat C-peptide.
- Size:200 μg
- Pk:200 µg
Mouse monoclonal antibody raised against synthetic peptide of C-peptide.
Type: Primary
Antigen: C-peptide
Clonality: Monoclonal
Clone: CII-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat