Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD8A Mouse Monoclonal Antibody [clone: MEM-31]
Anti-CD8A Mouse Monoclonal Antibody [clone: MEM-31]
Catalog # ABNOMAB3844
Supplier:  Abnova
Anti-CD8A Mouse Monoclonal Antibody [clone: MEM-31]
Catalog # ABNOMAB3844
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8 antigen, alpha chain
  • Antigen symbol:
    CD8A
  • Clonality:
    Monoclonal
  • Clone:
    MEM-31
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    925|926
  • Antigen synonyms:
    p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD8 from human crude thymus membrane fraction.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native CD8.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: MEM-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human