Anti-Disopyramide Monoclonal Antibody [clone: 11-433.5]
Catalog # ABNOMAB2220
Supplier: Abnova
Specifications
- Druh protilátky:Primární
- Název antigenu:Disopyramide
- Klonovalita:Monoclonal
- Klon:11-433.5
- Spojení:Unconjugated
- ELISA:Yes
- Izotyp:IgG1
- Křížová adsorbce:No
- Formát:Liquid
- Skladovací pufr:PBS, pH 7,4
- Skladovací teplota:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Imunogen:Disopyramide conjugated with KLH.
- Rozměry protilátky:100 μg
- Bal.:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against Disopyramide.
Type: Primary
Antigen: Disopyramide
Clonality: Monoclonal
Clone: 11-433.5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: