Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Disopyramide Monoclonal Antibody [clone: 13-788.1]
Anti-Disopyramide Monoclonal Antibody [clone: 13-788.1]
Catalog # ABNOMAB2222
Supplier:  Abnova
Anti-Disopyramide Monoclonal Antibody [clone: 13-788.1]
Catalog # ABNOMAB2222
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Disopyramide
  • Clonality:
    Monoclonal
  • Clone:
    13-788.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Isotype:
    IgG2b
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Disopyramide conjugated with KLH.
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against Disopyramide.

Type: Primary
Antigen: Disopyramide
Clonality: Monoclonal
Clone: 13-788.1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b
Reactivity: