Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Lidocaine Monoclonal Antibody [clone: 11-222.4]
Anti-Lidocaine Monoclonal Antibody [clone: 11-222.4]
Catalog # ABNOMAB2253
Supplier:  Abnova
Anti-Lidocaine Monoclonal Antibody [clone: 11-222.4]
Catalog # ABNOMAB2253
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lidocaine
  • Clonality:
    Monoclonal
  • Clone:
    11-222.4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Isotype:
    IgG1
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Lidocaine conjugated with KLH.
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against Lidocaine.

Type: Primary
Antigen: Lidocaine
Clonality: Monoclonal
Clone: 11-222.4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: