Anti-Procainamide Monoclonal Antibody [clone: 10-183.2]
Catalog # ABNOMAB2263
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Procainamide
- Clonality:Monoclonal
- Clone:10-183.2
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG2a
- Cross adsorption:No
- Format:Liquid
- Storage buffer:No additive
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Procainamide conjugated with KLH.
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against Procainamide.
Type: Primary
Antigen: Procainamide
Clonality: Monoclonal
Clone: 10-183.2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity: