Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-SHANK1 Mouse Monoclonal Antibody [clone: S22-21]
Catalog # ABNOMAB7892
Supplier:  Abnova
Anti-SHANK1 Mouse Monoclonal Antibody [clone: S22-21]
Catalog # ABNOMAB7892
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SH3 and multiple ankyrin repeat domains 1
  • Antigen symbol:
    SHANK1
  • Clonality:
    Monoclonal
  • Clone:
    S22-21
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    78957
  • Antigen synonyms:
    Sstrip|Spank1|Shank1a
  • Amino acid number:
    469 to 691
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 469-691 of rat Shank1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant Shank1.

Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml

Type: Primary
Antigen: SHANK1
Clonality: Monoclonal
Clone: S22-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat