Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GRIN2B Mouse Monoclonal Antibody [clone: S59-36]
Anti-GRIN2B Mouse Monoclonal Antibody [clone: S59-36]
Catalog # ABNOMAB7899
Supplier:  Abnova
Anti-GRIN2B Mouse Monoclonal Antibody [clone: S59-36]
Catalog # ABNOMAB7899
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate Ionotropic Receptor NMDA Type Subunit 2B
  • Antigen symbol:
    GRIN2B
  • Clonality:
    Monoclonal
  • Clone:
    S59-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    24410
  • Antigen synonyms:
    hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to N-terminus amino acids 20-271 of rat Grin2b.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant Grin2b.

Recommended Dilutions: Western Blot: 1-10 ?g/ml; Immunohistochemistry: 0.1-1.0 ?g/ml; Immunocytochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1-10 ?g/ml

Type: Primary
Antigen: GRIN2B
Clonality: Monoclonal
Clone: S59-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat