Anti-H2-D2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 28-14-8]
ABNOMAB5816
: Abnova
- Antibody type:Primary
- Antigen symbol:H2-D2
- Clonality:Monoclonal
- Clone:28-14-8
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from C3H.SW mouse splenocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native H2-D2.
Recommended Dilutions: Flow Cytometry: 0.5 ?g/10^6 cells
Type: Primary
Antigen: H2-D2
Clonality: Monoclonal
Clone: 28-14-8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse