Anti-H-2KK Mouse Monoclonal Antibody (Biotin) [clone: 16-3-22S]
Catalog # ABNOMAB5830
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1
- Antigen symbol:MHC1
- Clonality:Monoclonal
- Clone:16-3-22S
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:H2K1|H2K|H2KK|H-2 class I histocompatibility antigen K-K alpha chain
- Storage buffer:PBS (<
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from C3H.SW mouse splenocytes.
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against native H-2Kk.
Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells
Type: Primary
Antigen: H-2KK
Clonality: Monoclonal
Clone: 16-3-22S
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse