Anti-Bu-1a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 21-1A4]
ABNOMAB5844
: Abnova
- Antibody type:Primary
- Antigen symbol:Bu-1a
- Clonality:Monoclonal
- Clone:21-1A4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from Bursal cells of one-day-old H.B15 (Bu-1 a/b) strain chickens.
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against native Bu-1a.
Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells
Type: Primary
Antigen: Bu-1a
Clonality: Monoclonal
Clone: 21-1A4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken