Anti-Bu-1b Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5-11G2]
Catalog # ABNOMAB5846
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Bu-1b
- Clonality:Monoclonal
- Clone:5-11G2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native pruified from Bursal cells of 2- to 5-week-old CHA (Bu-1b) strain chickens.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native Bu-1b.
Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells
Type: Primary
Antigen: Bu-1b
Clonality: Monoclonal
Clone: 5-11G2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken