Anti-Bu-1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: AV20]
Catalog # ABNOMAB5853
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Bu-1
- Clonality:Monoclonal
- Clone:AV20
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from Bursal cells from one-day-old H.B15 (Bu-1a/b) strain chickens.
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against native Bu-1.
Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells
Type: Primary
Antigen: Bu-1
Clonality: Monoclonal
Clone: AV20
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken