Order Entry
Czech Republic
ContactUsLinkComponent
Anti-THY1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Anti-THY1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Catalog # ABNOMAB5958
Supplier:  Abnova
Anti-THY1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Catalog # ABNOMAB5958
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Thy1 Cell Surface Antigen
  • Antigen symbol:
    THY1
  • Clonality:
    Monoclonal
  • Clone:
    30-H12
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    21838
  • Antigen synonyms:
    Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
  • Storage buffer:
    PBS (1 mg/mL sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified Thy1 from mouse spleen cells or thymocyte membranes.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rat monoclonal antibody raised against native Thy1.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: 30-H12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse