Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Enterococcus species Rabbit Polyclonal Antibody
Anti-Enterococcus species Rabbit Polyclonal Antibody
  ABNOPAB13944
 :  Abnova
Anti-Enterococcus species Rabbit Polyclonal Antibody
  ABNOPAB13944
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Enterococcus species
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Bacteria
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    10 mM PBS, pH 7,2 (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Enterococcus faecium
  • Pk:
    1 mL

 

 

Rabbit polyclonal antibody raised against Enterococcus species.

Type: Primary
Antigen: Enterococcus species
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria