Order Entry
Czech Republic
ContactUsLinkComponent
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 6-13-19-1]
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 6-13-19-1]
Catalog # ABNOMAB5257
Supplier:  Abnova
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 6-13-19-1]
Catalog # ABNOMAB5257
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta-1,3-glucuronyltransferase 1
  • Antigen symbol:
    B3GAT1
  • Clonality:
    Monoclonal
  • Clone:
    6-13-19-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Tissue Culture Supernatant
  • Gene ID:
    27087
  • Antigen synonyms:
    LEU7|GLCATP|CD57|NK1|GLCUATP|NK-1|HNK1
  • Storage buffer:
    tissue culture supernatant
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified B3GAT1 from JURKAT T-cell line.
  • Size:
    1 mL
  • Pk:
    1 ml

Specifications

About this item

Mouse monoclonal antibody raised against native B3GAT1.

Recommended Dilutions: Immunofluorescence: 1:5

Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: 6-13-19-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human