Anti-PTPRC Mouse Monoclonal Antibody (Spectral Red®) [clone: F10-89-4]
ABNOMAB6068
: Abnova
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:F10-89-4
- Conjugation:Spectral Red®
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS (1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from T cells of human lymph nodes.
- Size:100 Reactions
- Pk:100 Tests
Mouse monoclonal antibody raised against native PTPRC.
Recommended Dilutions: Flow Cytometry: 10 ?l/10^6 cells
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: F10-89-4
Conjugation: Spectral Red®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human