Anti-CD4 Mouse Monoclonal Antibody [clone: ICO-86]
Catalog # ABNOMAB5408
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD4 molecule
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:ICO-86
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:920
- Antigen synonyms:L3T4|p55|CD4mut|Ly-4|W3/25
- Storage buffer:PBS, pH 7,4 (0,5% saccharose, 0,2% gelatin, 0,1% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:Native from T helper cells.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CD4.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: ICO-86
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human