Anti-SLC7A11 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # ABNOPAB14940
Supplier: Abnova
Specifications
- Antigen name:Solute Carrier Family 7 (Anionic Amino Acid Transporter Light Chain, Xc- System), Member 11
- Antigen symbol:Solute carrier family 7, member 11
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:26570
- Antigen synonyms:AI451155|xCT|sut|CCBR1
- Amino acid number:1 to 50
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 1-50 of mouse Slc7a11.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Slc7a11.
Type:
Antigen: SLC7A11
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human