Anti-ITGA5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5H10-27 (MFR5)]
Catalog # ABNOMAB9874
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 5
- Antigen symbol:ITGA5
- Clonality:Monoclonal
- Clone:5H10-27 (MFR5)
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:16402
- Antigen synonyms:FNRA|CD49e|Fibronectin Receptor Alpha Polypeptide|VLA-5|VLA5A
- Storage buffer:PBS (sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native Itga5 from mouse mast cell line MC/9.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rat monoclonal antibody raised against Itga5.
Recommended Dilutions: Flow Cytometry: <0.3 ?g/10^6 cells
Type: Primary
Antigen: ITGA5
Clonality: Monoclonal
Clone: 5H10-27 (MFR5)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse