Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CLIC1 Rabbit Polyclonal Antibody
Anti-CLIC1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18767
Supplier:  Abnova
Anti-CLIC1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18767
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chloride Intracellular Channel 1
  • Antigen symbol:
    CLIC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    1192
  • Antigen synonyms:
    Homo sapiens chloride intracellular channel 1 (CLIC1)|FLJ94671|CLIC1|Chloride intracellular channel 1 (cDNA|mRNA)
  • Storage buffer:
    buffer containing 0,02% sodium azide
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 14 amino acids at N-terminus of human CLIC1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of CLIC1.

Recommended Dilutions: ELISA: 1:20000-1:80000; Western Blot: 1:200-1:500; Immunohistochemistry: 1:50-1:250

Type: Primary
Antigen: CLIC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human