Anti-Os09g0482100 Mouse Monoclonal Antibody [clone: 31]
Catalog # ABNOMAB9953
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Os09g0482100
- Clonality:Monoclonal
- Clone:31
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rice
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4347402
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length Oryza sativa Os09g0482100.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant Os09g0482100.
Recommended Dilutions: Western Blot: 1:10000
Type: Primary
Antigen: Os09g0482100
Clonality: Monoclonal
Clone: 31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rice