Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-263]
Catalog # ABNOMAB9988
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 44 antigen
- Antigen symbol:CD44
- Clonality:Monoclonal
- Clone:MEM-263
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Monkey,Human,Dog
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- Storage buffer:PBS (15 mM sodium azide, 0,2% (w/v) BSA)
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
- Immunogen:COS-7 cells (African Green Monkey)
- Size:100 Reactions
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against CD44.
Recommended Dilutions: Flow Cytometry: 20 ?l reagent / 100 ?l of whole blood or 10^6 cells
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MEM-263
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Pig