Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC7A11 Rabbit Polyclonal Antibody
Anti-SLC7A11 Rabbit Polyclonal Antibody
Catalog # ABNOPAB8451
Supplier:  Abnova
Anti-SLC7A11 Rabbit Polyclonal Antibody
Catalog # ABNOPAB8451
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 7 (Anionic Amino Acid Transporter Light Chain, Xc- System), Member 11
  • Antigen symbol:
    Solute carrier family 7, member 11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    26570
  • Antigen synonyms:
    AI451155|xCT|sut|CCBR1
  • Storage buffer:
    PBS (0,1% proclin, 2.0% Block Ace)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to mouse Slc7a11.
  • Size:
    25 μg
  • Pk:
    25 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Slc7a11.

  • Recommended Dilutions: Immunohistochemistry: 5 μg/ml; Western Blot: 1 μg/ml

Type: Primary
Antigen: SLC7A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse