Anti-14-3-3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9657
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:14-3-3 protein
- Antigen symbol:14-3-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Zebrafish,Primates,Sheep,Chicken,Human,Bovine,Xenopus laevis,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:beta isoform|14-3-3 alpha|Protein 1054|Alpha polypeptide|Protein kinase C inhibitor protein 1|Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein|protein kinase C inhibitor protein-1|14-3-3 Protein beta/alpha|brain Protein 14-3-3
- Modification:phospho-Ser58
- Storage buffer:10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S58 of rat 14-3-3 protein.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of 14-3-3 protein.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: 14-3-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Clawed frog, Dog, Human, Mouse, Primates, Rat, Sheep, Zebrafish