Anti-CES1 Goat Polyclonal Antibody
Catalog # ABNOPAB7531
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carboxylesterase 1
- Antigen symbol:CES1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1066
- Antigen synonyms:CES2|CEH|carboxylesterase 2 (liver)|HMSE|carboxylesterase 1|SES1|triacylglycerol hydrolase|cholesteryl ester hydrolase|CES1|monocyte/macrophage serine-esterase 1|liver carboxylesterase|carboxylesterase 1 (monocyte/macrophage serine esterase 1)|MGC117365|ACAT|HMSE1|egasyn|acyl coenzyme A:cholesterol acyltransferase|TGH|PCE-1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-NTQAAQKLKDKE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human CES1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of CES1.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.03-0.1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-3 ?g/ml
Type: Primary
Antigen: CES1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human