Anti-17-beta-Estradiol-short linker Sheep Polyclonal Antibody
Catalog # ABNOPAB8209
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:17-beta-Estradiol-short linker
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Cross adsorption:No
- Format:Liquid
- Storage buffer:20 mM phosphate buffer, 150 mM NaCl, pH 7,2 (0,09% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:17 beta-Estradiol-short linker-BSA.
- Pk:1 mg
Specifications
About this item
Sheep polyclonal antibody raised against 17 beta-Estradiol-short linker.
Recommended Dilutions: Enzyme Immunoassay (3.5 ?g/ml
Type: Primary
Antigen: 17-beta-Estradiol-short linker
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: