About this item
The MEL-14 antibody is specific for mouse CD62L, also known as L-Selectin, a cell adhesion molecule which facilitates lymphocyte 'rolling' on activated vascular endothelium and homing to high endothelial venules (HEV) as immune cells transmigrate from blood into peripheral tissues. L-Selectin is a member of a family of Selectin molecules which act together with the integrin family of adhesion molecules to mediate leukocyte-endothelial interactions. L-Selectin is characteristically expressed by neutrophils, and is also found on B cells, monocytes, granulocytes, and at varying levels on naive, effector and memory T cells. It is rapidly shed upon cell activation, releasing into the circulation a soluble form whose biological role is of particular interest in cancer biology research.
The MEL-14 antibody may be used as a phenotypic marker for CD62L expression on a variety of immune cell types. Please note that CD62L (L-Selectin) itself is also referred to as MEL-14 in the literature.
Type: Primary
Antigen: CD62L (L-Selectin)
Clonality: Monoclonal
Clone: MEL-14
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Specifications
- Catalog No:
- 80-0621-U025
- 80-0621-U100
- Antigen symbol:
- SELL
- SELL
- Antigen name:
- Selectin L
- Selectin L
- Antigen synonyms:
- LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- redFluor® 710
- redFluor® 710
- Clone:
- MEL-14
- MEL-14
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 20343
- 20343
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- 2...8 °C, protected from light
- 2...8 °C, protected from light
- Tested applications:
- Flow Cytometry
- Flow Cytometry