Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-2A5]
Catalog # 70-7102-U100
Supplier:  Tonbo Biosciences
Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-2A5]
Catalog # 70-7102-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 10
  • Antigen symbol:
    IL10
  • Clonality:
    Monoclonal
  • Clone:
    JES5-2A5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    16153
  • Antigen synonyms:
    IL10A|IL-10|GVHDS|TGIF|CSIF
  • Storage temperature:
    +2 °C to +8 °C
  • Pk:
    100 µG

Specifications

About this item

The JES5-2A5 antibody is specific for mouse IL-10. IL-10 has numerous biological activities including the ability to inhibit both the synthesis of Th1 cytokines and proliferation of T cells. In the mouse, IL-10 is produced by phagocytic cells, conventional dendritic, cells, T cells, B cells, and NK cells. The JES5-2A5 antibody is a neutralizing antibody.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES5-2A5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse