Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IL-4 Rat Monoclonal Antibody (In Vivo Ready™) [clone: MP4-25D2]
Anti-IL-4 Rat Monoclonal Antibody (In Vivo Ready™) [clone: MP4-25D2]
Supplier:  Tonbo Biosciences
Certificates
Anti-IL-4 Rat Monoclonal Antibody (In Vivo Ready™) [clone: MP4-25D2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 40-7048-U100
  • 40-7048-U500
  • Antigen symbol:
  • IL4
  • IL4
  • Antigen name:
  • Interleukin 4
  • Interleukin 4
  • Antigen synonyms:
  • BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
  • BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • MP4-25D2
  • MP4-25D2
  • Reactivity:
  • Rhesus monkey, Human
  • Rhesus monkey, Human
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 3565
  • 3565
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C
  • +2 °C to +8 °C

Specifications

Related Information

Frequently Bought Together