Order Entry
Czech Republic
ContactUsLinkComponent
Rat IgG1 Isotype Control [clone: HRPN]
Rat IgG1 Isotype Control [clone: HRPN]
Catalog # 70-4301-U100
Supplier:  Tonbo Biosciences
Rat IgG1 Isotype Control [clone: HRPN]
Catalog # 70-4301-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clone:
    HRPN
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Storage temperature:
    Store Undiluted at 2 to 8 °C
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: N/A