Anti-CD178 (Fas ligand) Mouse Antibody (Biotin) [clone: NOK-1]
Type:
Antigen: CD178 (Fas ligand)
Clonality:
Clone: NOK-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- 30-9919-U025
- 30-9919-U100
- Antigen symbol:
- FASLG
- FASLG
- Antigen name:
- Fas Ligand
- Fas Ligand
- Antigen synonyms:
- APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
- APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
- Antibody type:
- Primary
- Conjugation:
- Biotin
- Biotin
- Clone:
- NOK-1
- NOK-1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 356
- 356
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
Frequently Bought Together