Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD178 (Fas ligand) Mouse Antibody (In Vivo Ready™) [clone: NOK-1]
Anti-CD178 (Fas ligand) Mouse Antibody (In Vivo Ready™) [clone: NOK-1]
  40-9919-U100
Anti-CD178 (Fas ligand) Mouse Antibody (In Vivo Ready™) [clone: NOK-1]
  40-9919-U100

 

  • Antibody type:
    Primary
  • Antigen name:
    Fas Ligand
  • Antigen symbol:
    FASLG
  • Clone:
    NOK-1
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Neutralisation:
    Yes
  • Gene ID:
    356
  • Antigen synonyms:
    APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
  • Storage temperature:
    Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

The NOK-1 antibody reacts with human CD178 (Fas ligand) in both membrane bound and soluble forms. Fas ligand is a 40 kDa transmembrane glycoprotein, a member of the TNF family, and is expressed by activated T and NK cells, neutrophils, and monocytes. Interactions between CD178 (Fas ligand) and CD95 (Fas) induce a program of apoptosis and play a key role in immune regulation and homeostasis. The extracellular domain of human CD178 can be cleaved from the surface by matrix metalloproteinases (MMPs) resulting in a 26 kDa soluble protein.

Type: Primary
Antigen: CD178 (Fas ligand)
Clonality:
Clone: NOK-1
Conjugation: In Vivo Ready™
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human