Anti-CD154 Armenian Hamster Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MR1]
Supplier: Tonbo Biosciences
Certificates
About this item
The MR1 antibody is specific for mouse CD154, a 39 kD glycoprotein that is primarily expressed on activated T cells. CD154 (CD40 ligand, gp39) binds to CD40 expressed on antigen presenting cells and mediates important costimulatory functions between these cells. Signals generated via cognate interactions between CD154-CD40 on activated T and B cells are required for T-dependent B cell responses. The MR1 antibody is reported to block interaction of CD154 with CD40.
Type:
Antigen: CD154
Clonality:
Clone: MR1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 35-1541-U025
- 35-1541-U100
- Antigen symbol:
- CD40L
- CD40L
- Antigen name:
- CD40 Ligand
- CD40 Ligand
- Antigen synonyms:
- hCD40L|TRAP|CD40L|IMD3|gp39|CD154|IGM|T-BAM|TNFSF5|HIGM1|CD40 ligand
- hCD40L|TRAP|CD40L|IMD3|gp39|CD154|IGM|T-BAM|TNFSF5|HIGM1|CD40 ligand
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- MR1
- MR1
- Reactivity:
- Mouse
- Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 21947
- 21947
- Isotype:
- IgG
- IgG
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2-8 °C, protect from light, do not freeze.
- Store undiluted at 2-8 °C, protect from light, do not freeze.
Specifications
Frequently Bought Together